Digi-Sense Prec. Ind. PRT, 0.187" x 9" , -60~160C, 5-Pin Din
Representative image only.
Digi-Sense Item # WW-90452-07
State-of-the-art, high-accuracy temperature probes at a fraction of the cost
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypePrecision Industrial PRT/RTD
  • Min Temperature (° F)-76
  • Max Temperature (° F)320
  • Min Temperature (° C)-60
  • Max Temperature (° C)160
  • Temperature Accuracy±0.05°C and ±0.04°C/100 hr
  • Response Time (Sec)4
  • Connector Type5-Pin DIN
  • Sheath Length (in)9
  • Sheath Diameter (in)0.1875
  • DescriptionPrec. Ind. PRT, 0.187\" x 9\" , -60~160C, 5-Pin Din
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
AM1610 Series—Precision industrial PRT/RTD probes: Conform to the standard 385 curve, letting you use your DIN/IEC RTD meters fully. Manufactured using a special aged and tested 5mm long sensing element to achieve the best performance in stability and repeatability. All have an accuracy of 35 mK at 0°C and short-term stability of 10 mK at 0°C. The AM1610 has a temperature range of -60 to 160ºC (-76 to 320ºF)