Digi-Sense Mechanical/Winding Stopwatch, White-Face; 30 min
Representative image only.
Digi-Sense Item # WW-98768-50
Simple to use—single-action stopwatch only has one button to press
  • Start, stop and return to zero by consecutive crown depressions
  • Times up to 30 minutes with 0.01-second resolution
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Timing Capacity30 minutes
  • Count DirectionUp
  • Number of Channels1
  • Resolution1/100 second
  • CaseBrass with chromium plating
  • LensABS plastic
  • Outside Diameter (OD) (in)2
  • DescriptionMechanical/Winding Stopwatch, White-Face; 30 min
  • Warranty1 year
1 Year Warranty
Key Features
  • Start, stop and return to zero by consecutive crown depressions
  • Times up to 30 minutes with 0.01-second resolution