Digi-Sense Mason Hygrometer with 2 Spirit-Filled Glass Thermometers
Representative image only.
Digi-Sense Item # WW-03311-02
Calculate relative humidity with wet and dry bulb temperatures
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)0
  • Min Temperature (° C)-15
  • Max Temperature (° F)120
  • Max Temperature (° C)50
  • Height (in)12 1/4
  • Width (in)12 1/4
  • Depth (in)3/4
  • DescriptionMason Hygrometer with 2 Spirit-Filled Glass Thermometers
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Mason hygrometer features two red spirit filled thermometers contained within a sturdy plastic case. Included chart converts wet and dry bulb temperatures to humidity values. Dual temperature scales range from 0 to 120°F (-15 to 50°C).