Digi-Sense K30-2-506 Thermocouple Wire Type K 30-Gauge, FEP Insulation, 100-ft Spool
Representative image only.
Cole-Parmer Item # WW-08541-02
Make your own extension cables
  • 6 month warranty
PRODUCT OPTIONS
₹13,600.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialFluorinated ethylene propylene (FEP)
  • Wire Gauge (Ga)30
  • Error Limit±2% reading from -330 to -165°F (-200 to -110°C); ±4°F (±2.2°C) from -165 to 545°F (-110 to 285°C); ±0.75% reading from 545 to 2300°F (285 to 1250°C)
  • Inner InsulationFluorinated ethylene propylene (FEP)
  • Outer InsulationFluorinated ethylene propylene (FEP)
  • Stranded / SolidSolid
  • Special InformationSPECIAL LIMITS OF ERROR
  • Min Temperature (° F)-328
  • Max Temperature (° F)400
  • Min Temperature (° C)-200
  • Max Temperature (° C)204
  • Length (ft)100
  • Size (in)0.030 in x 0.048 in
  • DescriptionThermocouple Wire Type K 30-Gauge, FEP Insulation, 100-ft Spool
6 Month Warranty
Key Features
  • 6 month warranty
More About this Item
Wire comes in 30 gauge size, ideal for fabricating your own probes or extension cables (meets ANSI and ASTM standards). Choose from wire with fluorinated ethylene propylene resin (FEP/FEP) or fiberglass materials as well as in 100-ft or 1000-ft lengths for any application.