Digi-Sense Infrared Thermometer with Type-K Input
Representative image only.
Digi-Sense Item # WW-39644-00
Measure both noncontact and contact temperature
  • Display infrared and contact temperatures simultaneously
  • Laser sighting and adjustable emissivity
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Distance To Target Size Ratio (D:S)12:1
  • EmissivityAdjustable
  • Laser ClassClass II
  • Min Temperature (° F)-76
  • Max Temperature (° F)932
  • Min Temperature (° C)-60
  • Max Temperature (° C)500
  • AccuracyInfrared: ±2% of reading or ±3.6°F, whichever is greater; Contact: ±1% of reading or ±1.8°F, whichever is greater (Infrared: ±2% of reading or ±2.0°C, whichever is greater; Contact: ±1% or ±1.0°C, whichever is greater)
  • Response time<1 second (90% response)
  • Low Battery IndicatorYes
  • BatteryAAA Battery (2)
  • DescriptionInfrared Thermometer with Type-K Input
  • Warranty1 Year
1 Year WarrantyConformite EuropeenneLaser Class IILimited Lifetime Warranty
Key Features
  • Display infrared and contact temperatures simultaneously
  • Laser sighting and adjustable emissivity
  • 1 year warranty
More About this Item
These infrared thermometers provide precision spot temperature measurement in a wide range of applications. Ideal for use in the laboratory environment, food preparation areas, plant maintenance, automotive repair, HVAC testing, electrical troubleshooting, and many others. This series of thermometers feature advanced optics that allows the measuring of smaller objects from a longer distance. Offering a greater degree of measuring flexibility then other products with adjustable emissivity, min/max and average, min/max differential, lock function for continuous measurement, high and low audible and visual alarm limits, automatic data hold, and auto off. Includes a built-in thermocouple port that will accommodate any type-K thermocouple with a mini-connector (thermocouple sold separately), when direct contact measurement is required. Adjustable emissivity provides for greater measuring capabilities on a wider range of surface. The laser and LCD backlight have the capability of being turned on or off independently of each other, depending on the requirements of the applications. The LCD simultaneously displays the current temperature in °F or °C, the pre-selected emissivity, the laser scan, and battery life icons.
The optional belt-mountable case lets you conveniently carry around your infrared thermometer-sold separately below.
What's Included: two AAA batteries.