Digi-Sense Industrial RTD Probe 4" Length, 1/2" Thread
Representative image only.
Digi-Sense Item # WW-93820-40
All-in-one, includes industrial probe head, element and thermowell
  • 1 year warranty
PRODUCT OPTIONS
₹42,600.00INR / Each
₹27,000.00 - ₹33,700.00INR / Each
for an additional
₹27,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeRTD
  • Length (\")4
  • Min Temperature (° F)-50
  • Max Temperature (° F)350
  • Min Temperature (° C)-45
  • Max Temperature (° C)176
  • JunctionUngrounded
  • Sheath Length (in)4
  • Sheath Diameter (in)0.5
  • DescriptionIndustrial RTD Probe 4\" Length, 1/2\" Thread
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Industrial RTDs assemblies produce greater accuracy are ideal for use in industrial applications, chemical processes, production lines, ovens, furnaces and vessels. . The probes conductor is made of 24 gauge cooper wire and is encased with FEP insulation, and has a 316 stainless steel sheath. Featuring a standard size protection head with a gasketed cap to seal out dust and moisture and houses a ceramic terminal block. Head has a 3/4" NPT (F) opening on the side for easy connection to electrical conduit. 316 stainless steel thermowells are included in assemble and are ideal for applications where removing of the probe without draining the vessel is necessary.