Digi-Sense Hands-Free Kit for Temperature Meters
Representative image only.
Digi-Sense Item # WW-35427-85
  • 3-way hands-free operation
  • Stand it
  • Stick it
  • Hang it
PRODUCT OPTIONS
₹7,600.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionHands-Free Kit for Temperature Meters
Key Features
  • 3-way hands-free operation
  • Stand it
  • Stick it
  • Hang it
More About this Item
Get the most out of your thermocouple meter with these accessories.
OAKTON’s exclusive “3-WAY HANDS-FREE” capability allows “hands-free” use—use the built-in stand on a table top, attach the meter to metal objects through magnets inserted into the armor boot, or hang from a pipe or belt using the hook-and-loop strap.