Accuracy±1% of reading + 1.8⁰F (1⁰C) below -148⁰F (-99.9⁰C); ±1% of reading 0.9⁰F (0.5⁰C) above -148⁰F (-99.9⁰C)
Resolution0.1⁰F (0.1⁰C) from –148 to 1832⁰F, (-99.9 to 999.9⁰C); 1⁰F (1⁰C) outside range
Data LoggingYes
Memory Capacity16000 data points
AlarmYes
Low Battery IndicatorYes
Display TypeLarge, dual, backlit LCD
Battery Life3000 hours
BatteryOne 9V battery (included)
DescriptionDual-Input Data Logging Thermocouple Thermometer System with NIST-Traceable Calibration
Includes
Two general-purpose Type-K thermocouple probes, protective rubber boot with built-in stand, one 9 V battery, a USB cable, data logging software, and NIST-traceable calibration report with data supplied by InnoCal.
Key Features
Reliable measurements, guaranteed!
Choose from meter or meter system—both include calibration
Heavy duty construction to protect against knocks and jars.
Super large LCD for easy viewing of data
Min, Max, and Avg readings, for quick analysis of the object
Electronic Offset function allows compensation of probe errors
Bundled kit for convenience and savings
Two (2) probes and NIST-Traceable calibration included!
Record up to 16,000 measurements for expanded data analysis capability
Software provides quick and easy data interpretation
Temperature differential plus Min, Max, and Avg readings, for quick analysis
Relative time clock on Min, Max, and Avg provides a time reference for major events
USB Interface
Accepts a wide variety of other probes
2 year warranty
More About this Item
The Digi-Sense Professional Thermocouple Thermometers offer fast response and laboratory accuracy. Features include automatic backlight, Max/Min readings, and automatic power-off. These thermometers provide precise measurements when accuracy is a must, delivering dependable results every time! Each model is packed with distinctive features for use in a wide array of applications. Robust and reliable constructed to protects against knocks and jars. These thermometers produce a high degree of accuracy, with a rapid response time, and exceptional battery life of up to 3000 hours.
Digi-Sense instrument are fully tested and calibrated to NIST-traceable standards, saving you hundreds of dollars and extra waiting time to calibrate separately.
Advanced features include data logging, USB interface, relative time clock, offset compensation, temperature differential.