Cole-Parmer Essentials Dry Block/Incubator Liquid-In-Glass Thermometer; 0 to 80C, 100mm Immersion, Organic Liquid Fill

Representative image only.
Cole-Parmer Essentials Item # WW-08006-19
Replace mercury thermometers for a specific application with the same accuracy without any of the hazards
  • Non-mercury liquid
  • 5 year warranty
Monitor your critical environments

SPECS & DESCRIPTION

Specifications & Description

  • Length (\")8 29/32
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • Accuracy±0.5°C
  • Division (° C)0.5
  • Thermometer FillRed Spirit
  • Overall Length (in)8.9
  • Overall Length (mm)225
  • Immersion Depth (mm)100
  • Stem MaterialWhite-Back Glass
  • DescriptionDry Block/Incubator Liquid-In-Glass Thermometer; 0 to 80C, 100mm Immersion, Organic Liquid Fill
  • Warranty5 years
5 Year Warranty

Key Features

  • Non-mercury liquid
  • 5 year warranty

More About this Item

These thermometers are designed for use in specific applications. These Durac thermometers feature markings that are permanently printed on the glass, individually serialized, and red spirit liquid that has a dark scale contrasts against a white background. Some thermometers are PFA coated for added safety which may cause a slight change in accuracy up to 1 division.

Dry block thermometers - designed for use in all standard heating blocks and water baths with most having arrows indicating critical temperatures.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat