This item is now discontinued. We recommend the following as suggested replacements: WW-92000-02
Digi-Sense 12 Channel Scanning Data Logging Benchtop Thermocouple Thermometer
Representative image only.
Digi-Sense Item # WW-92000-01
No need to juggle twelve different data logging thermometers
  • Each channel is independently programmable with thermocouple type and alarm conditions
  • Large full-color LCD displays each channel simultaneously
  • Log up to 296,296 data points and download or stream to PC
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)-328
  • Max Temperature (° F)2502
  • Min Temperature (° C)-200
  • Max Temperature (° C)1372
  • Accuracy0.1% Span
  • Resolution0.1˚C
  • Output24 V, 1 Amp
  • Data LoggingYes
  • AlarmAudible/Visual
  • Display TypeLCD
  • Height (in)4
  • Width (in)6 1/2
  • Length (in)6
  • Description12 Channel Scanning Data Logging Benchtop Thermocouple Thermometer
  • Warranty2 year
Two year warranty
Key Features
  • Each channel is independently programmable with thermocouple type and alarm conditions
  • Large full-color LCD displays each channel simultaneously
  • Log up to 296,296 data points and download or stream to PC
  • 2 year warranty
More About this Item
This 12-channel scanning thermometer simultaneously monitors different temperature processes and provides real-time data on your PC with the supplied data acquisition software. The thermometer also functions as a data logger, storing readings in internal nonvolatile memory for later download to PC in CSV format.
Audible and visible alarms alert you when a channel logs a temperature exceeding the high or low set point. Thermocouple temperature probes are sold separately.