Dickson 8" Circular Chart Recorder Paper, -50 to 50°F/C, 31 Days, 60/Pack
Representative image only.
Dickson Mfr # C406 – Item # WW-80006-70
Monitor with precision accuracy for quality control
  • Only authentic Dickson charts and pens guarantee accuracy
  • Dickson charts are a great way to archive your monitoring history
  • Flexible options to choose time, temperature, and size
PRODUCT OPTIONS
₹14,400.00INR / Pkg of 60
SPECS & DESCRIPTION
Specifications & Description
  • Chart TypeCircular
  • Chart Speed31 day
  • Chart Diameter (in)8
  • Max Temperature (° F)50
  • Min Temperature (° F)-50
  • Max Temperature (° C)50
  • Min Temperature (° C)-50
  • Description8\" Circular Chart Recorder Paper, -50 to 50°F/C, 31 Days, 60/Pack
Key Features
  • Only authentic Dickson charts and pens guarantee accuracy
  • Dickson charts are a great way to archive your monitoring history
  • Flexible options to choose time, temperature, and size
More About this Item
Choose the paper to meet your application needs.