Diba Omnifit® PTFE Cone (Ferrule) for Connector and Valve Caps, 1.0 mm; 4/pk
Representative image only.
Diba Mfr # 001507 – Item # WW-21939-50
Select the size you need for a leaktight seal
  • Chemically resistant PTFE construction
SPECS & DESCRIPTION
Specifications & Description
  • Compression Size / Tube Od (Port 1) (mm)1
  • DescriptionOmnifit® PTFE Cone (Ferrule) for Connector and Valve Caps, 1.0 mm; 4/pk
Key Features
  • Chemically resistant PTFE construction
More About this Item
Select PTFE cones when chemical resistance is important. The OD of the tubing should be matched to the inside diameter of the cone.