Diba Omnifit® High Flow Bubble Trap, PVC, 10 µm PTFE filter; 1/ea
Representative image only.
Diba Mfr # 006BT-HF – Item # WW-21940-39
Remove bubbles with or without vacuum assistance
  • Constructed with durable PTFE membrane
PRODUCT OPTIONS
₹13,300.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPVC body, PTFE membrane
  • Flow Rate60 mL/min (max)
  • Port Size1/4\"-28 UNF(F)
  • Max Pressure (PSI)30
Key Features
  • Constructed with durable PTFE membrane
More About this Item
Effectively remove bubbles* from aqueous solutions inline or downstream with bubble traps. Traps feature ¼"-28 UNF(F) port and come with 10 µm PTFE filter. The high-flow trap features a vacuum port on the atmosphere (dry) side of the membrane. When vacuum is applied, the pressure differential between the system and atmosphere side increases (must not exceed 30 psi/2.1 bar), causing bubbles to be removed from the liquid and permitting increased liquid flow.
*These bubble traps do not remove dissolved gases.