Diba Omnifit® Bottom-of-Bottle Filter for GL32 /38-430 Bottle, 10 µm PE; 1/ea
Representative image only.
Diba Mfr # 009F-32-10-PE – Item # WW-21943-33
Push-fit onto a 1/8" OD tube for a quick connection
  • Optimize solvent filtration with filters and bubblers
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialPolyethylene
  • Pore Size (micron)10 µm
  • Housing MaterialPTFE
  • ConnectionsFor GL32/38-430 bottle
  • Ports1/8\" OD tube connection
Key Features
  • Optimize solvent filtration with filters and bubblers
More About this Item
These reservoir filters and bubbler provide simple and effective solvent filtration. A 10 µm filter ensures even the smallest particulates are removed from the solvent supply line. Bottom-of-bottle filters have slots in the body allowing virtually all liquid to be removed from the bottle, minimizing waste. Select from PTFE body/membrane or PTFE body/PE membrane filters. Filter bubbler, suitable for filtration or sparging applications, comes with a 316 SS filter.