Diba 1001 Standard fittings, two PP caps, four large O-rings, 1 to 4 mm OD, 3/pack
Representative image only.
Diba Mfr # 001001 – Item # WW-06473-00
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPTFE
  • Connection TypeHPLC/Chromatography
  • Compression Size / Tube Od (Port 1) (mm)4
  • Compression Size / Tube Od (Port 2) (mm)4
  • DescriptionStandard fittings, two PP caps, four large O-rings, 1 to 4 mm OD, 3/pack
More About this Item
All fittings include polypropylene caps, 2 large FKM (fluoroelastomer) O-rings, 1 small FKM O-ring, and 1 PTFE cone. Connector bodies are PTFE. Additional seals/cones available for separate purchase; see the accessories listing for this product line for more information.