DeFelsko PosiTector 200C1 Coating Thickness Gauge, Non-Metals, 2-150 mils
Representative image only.
DeFelsko Mfr # 200C1-E – Item # WW-59784-04
Quickly measure coating thickness on all non-metal substrates
  • Ready to measure - no calibration adjustment required to measure most coatings
  • Solvent, acid, oil, water and dust resistant - meets or exceeds IP5X
  • Internal memory for 250 readings in a single batch to be view or downloaded
  • Statistics mode continuously displays average, standard deviation, and number of readings while measuring
  • High contrast backlit display for bright or dark environments
  • Certificate of calibration showing traceability to NIST or PTB included (Long Form)
  • Proven non-destructive ultrasonic technique conforms to ASTM6132 and ISO2808
  • Selectable display languages
  • Non-destructively measures thickness of coating 2 to 150 mils (50 to 3800 microns)
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Range2 to 150 mil
  • DisplayLCD
  • DescriptionCoating Thickness Gauge, Non-Metals, 2-150 mils
Two year warranty
Key Features
  • Ready to measure - no calibration adjustment required to measure most coatings
  • Solvent, acid, oil, water and dust resistant - meets or exceeds IP5X
  • Internal memory for 250 readings in a single batch to be view or downloaded
  • Statistics mode continuously displays average, standard deviation, and number of readings while measuring
  • High contrast backlit display for bright or dark environments
  • Certificate of calibration showing traceability to NIST or PTB included (Long Form)
  • Proven non-destructive ultrasonic technique conforms to ASTM6132 and ISO2808
  • Selectable display languages
  • Non-destructively measures thickness of coating 2 to 150 mils (50 to 3800 microns)
  • 2 year warranty
More About this Item
The standard ultrasonic coating thickness gauges non-destructively measure the thickness of coatings on non-metal substrates including wood, plastic, composites, and more. They also measure the total thickness of a coating system.