Decon Labs CiDecon 8511 Plus Wipes; 180 Wipes/Canister, 12 Canister/Case
Representative image only.
Decon Labs Mfr # 8511 – Item # WW-86115-58
Provides thorough disinfection efficacy
  • Effective against broad range of pathogenic organisms including TB
  • FDA compliant for use on medical equipment and instruments
  • Meets OSHA’s Blood Borne Pathogens Standard
  • EPA registered
₹76,500.00INR / Case of 1
SPECS & DESCRIPTION
Specifications & Description
  • FormatWipes
  • DescriptionPlus Wipes; 180 Wipes/Tub, 12 Tubs/Case
Key Features
  • Effective against broad range of pathogenic organisms including TB
  • FDA compliant for use on medical equipment and instruments
  • Meets OSHA’s Blood Borne Pathogens Standard
  • EPA registered
More About this Item
Pre-moistened hard surface disinfectant wipes are ideal for use in labs, hospitals, and anywhere disinfectant efficacy is required. CiDecon Plus Wipes, which come in a convenient “pop-up” canister, are economical, durable, and display superior efficacy against a broad range of pathogenic organisms. Achieves disinfection in 10 minutes and maintains continual residual bacteriostatic activity for up to six months.
Not available in Canada.