Data-Logger for Traceable® Dual-Input/Output Thermocouple Thermometer
Representative image only.
Item # WW-98768-09
Record data—even if you're away from your PC
  • For use with Traceable® meters featuring computer output
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible With91210-03
  • DescriptionData-Logger for 91210-03
  • Warranty1 Year
1 Year WarrantyNIST Traceable Calibration
Key Features
  • For use with Traceable® meters featuring computer output
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • 1 year warranty
More About this Item
This data logger is complete DAS–4™ System captures and stores up to 8000 bytes—over 1000 readings. The readings may be taken at intervals from 1 second to 99 hours.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.