CPC (Colder) MPC17004T39 Quick-Disconnect Body, PS, 1/4" ID; 5/Pk
Representative image only.
CPC (Colder) Mfr # MPC17004T39 – Item # WW-31311-21
₹2,600.00INR / Pkg of 5
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolysulfone
  • Sterilization MethodAutoclave, ethylene oxide (ETO), or gamma
  • Connection TypeQuick-Disconnect x Barb
  • Barb Size / Tubing Id (Port 1) (in)1/4
  • Min Temperature (° F)-40
  • Max Temperature (° F)300
  • Min Temperature (° C)-40
  • Max Temperature (° C)148
  • Max Operating Pressure (PSI)120
  • Max Vacuum (in Hg)25
  • DescriptionQuick-disconnect fittings, Polysulfone fittings, 1/4" ID, body, 5/pack
Certificate of Compliance
More About this Item
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).