Corning SLP ThermalTray Thermoconductive Platform, slim, low profile; 1/ea
Representative image only.
Corning Mfr # 432073 – Item # WW-04392-73
Don't let your samples sinkDon't let your samples sink
  • Support containers and tube racks in liquid temperature-control media
  • Highly conductive alloy is compatible with liquid nitrogen
  • Sterilize by autoclave or dry heat
  • 1 year warranty
PRODUCT OPTIONS
₹93,200.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")11
  • Width (InchShort)5 1/2
  • Height (in)1 1/4
  • MaterialThermoconductive Alloy
  • ColorGray
  • Compatible WithLiquid nitrogen
  • DescriptionThermoconductive Platform, slim, low profile; 1/ea
  • Warranty1 year
1 Year Warranty
Key Features
  • Support containers and tube racks in liquid temperature-control media
  • Highly conductive alloy is compatible with liquid nitrogen
  • Sterilize by autoclave or dry heat
  • 1 year warranty
More About this Item
These thermally conductive trays support samples in water baths, melting ice, and liquid nitrogen, keeping the sample out of the liquid environment. Alloy construction is sturdy, stable, and ensures a uniform temperature across entire surface. Resistant to bleach, alcohol, other disinfectants, and laboratory detergents.