Corning LX55 CoolSink, each

Representative image only.
Corning Mfr # 432072 – Item # WW-04392-72
Rapid temperature adaption to any temperature source
  • Reduce temperature variability and uncertainty with uniform well-to-well temperature
  • Ensured temperature accuracy of ±0.1° C across all tubes and plates for critical applications
  • Easy to clean or sterilize
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Lowest Set Temperature (° C)-196
  • Height (in)1 2/5
  • Length (in)5 3/4
  • Width (in)2 1/2
  • Length (cm)14.6
  • Height (cm)3.5
  • Width (cm)6.4
  • CapacityOne 55-mL reagent reservoir
  • DescriptionCoolSink LX55
  • Warranty1 year
1 Year Warranty

Key Features

  • Reduce temperature variability and uncertainty with uniform well-to-well temperature
  • Ensured temperature accuracy of ±0.1° C across all tubes and plates for critical applications
  • Easy to clean or sterilize
  • 1 year warranty

More About this Item

These thermo-conductive plate and reservoir modules provide uniform temperature to all wells when cooling, snap freezing, heating, or thawing samples. Ideal for use in a variety of temperature sources such as ice, dry ice, liquid nitrogen, or water baths. The modules will rapidly adapt to the temperature of the source being used, from -196°C to >100°C. All options may be autoclaved, high heat sterilized, or decontaminated with bleach, alcohol, or other disinfectants or lab detergents. Both plate modules are SBS-compatible.
Use in traditional ice buckets and pans, or for an ice-free option, use with Corning CoolBox ice-free cooling systems.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat