Corning Gosselin Petri Dish, Polystyrene, Clean Packaging, Stackable, Sterile, 3 Vents, 100 x 15 mm; 825/Cs
Representative image only.
Corning Gosselin Mfr # BP93B-102 – Item # WW-34551-83
Optimal flatness ensures constant depth of agar
  • Vented dishes reduce condensation and allow air circulation
  • Class 100 manufacturing environment ensures aseptic production and packaging
  • Packed 33 per Sleeve, 25 Sleeve per Case
PRODUCT OPTIONS
₹29,800.00INR / Case of 825
Parafilm PM996 M Wrap, 4
₹3,800.00INR / Roll of 1
Consider Always in Stock Alternative
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolystyrene
  • Closure3 Vents; Stackable
  • Height (mm)15
  • Diameter (mm)100
  • SterileSterile
  • PackagingClean Packaging
  • DescriptionPetri Dish, Polystyrene, Clean Packaging, Stackable, Sterile, 3 Vents, 100 x 15 mm; 825/Cs
Key Features
  • Vented dishes reduce condensation and allow air circulation
  • Class 100 manufacturing environment ensures aseptic production and packaging
  • Packed 33 per Sleeve, 25 Sleeve per Case
More About this Item
These petri dishes are constructed of crystal polystyrene with an injection point located on the side of the dish bottom to optimize clarity even by image analyzing systems. Dishes are available in slippable or stackable designs, sterile or aseptic, and general lab or traceable clean packaging.
Slippable dishes are automation-compatible while stackable dishes have a stacking ring which makes them practical to pile.
Dishes are available in aseptic and sterile versions. Aseptic products are manufactured in a controlled environment, without human intervention during the manufacturing process. Sterile products are sterilized using an irradiation method compliant with the requirements of ANSI/AAMI/ISO 11137 to achieve (SAL 10⁻⁶).
General lab packaging does not have an outer bag, making it more economic and environmentally friendly. Clean packaging features two outer bags and bar code label for optimal traceability and suitable for clean room use.