Corning Gosselin S-Blender-1A Paddle Blender, solid door, adjustable settings, 80 to 400 mL; 115 VAC
Representative image only.
Corning Gosselin Mfr # S-Blender-1A – Item # WW-34551-08
Variable settings for quick and exact sample blending
  • Adjust time, speed, and blending strength to your unique application
  • Simple to use and read digital control, keypad, and LCD
  • Easy to clean and disassemble with autoclavable paddles and door
SPECS & DESCRIPTION
Specifications & Description
  • Bag Capacity (mL)400
  • Min Blending Speed (strokes)450
  • Max Blending Speed (strokes)650
  • Power (VAC)115
  • Door TypeFront loading, stainless steel
  • Timer (minutes)30 to 210 seconds or continous mode
  • DescriptionBlender, Adjustable, 80 to 400 mL
3 Year Warranty
Key Features
  • Adjust time, speed, and blending strength to your unique application
  • Simple to use and read digital control, keypad, and LCD
  • Easy to clean and disassemble with autoclavable paddles and door
More About this Item
The Gosselin blender uses paddles to homogenize sample dilutions inside PE bag sizes from 80 mL to 400 mL. The paddles crush and extract organisms into suspension. You can adjust time, speed, and blending strength for a suitable blending of your samples. Adjust the LCD timer to run between 30 seconds to 210 seconds, or if you prefer, you can run the blender continuously. The variable speed starts at a slow 450 strokes per minute, then a medium 550, and up to a fast 650 strokes per minute. For a quick setup, program nine customized blending modes.
The blender runs quietly like a humming refrigerator with a motor loudness at just 51 dBA. The motor is protected with an electronic circuit breaker, and the blender’s door stays closed during operation.
For cleaning the blender, the sample chamber is removable, and the stainless steel door and paddles are autoclavable. Order sterile PE bags and clips separately.