This item is now discontinued. We recommend the following as suggested replacements: WW-34551-70
Corning Gosselin Round Bottle, 100 mL, HDPE, 37 mm red screw cap, sterile; 145/cs
Representative image only.
Corning Gosselin Mfr # LR100-12 – Item # WW-34551-99
Ideal lightweight and shockproof bottle for media sampling and storage
  • Wide mouth with red screw cap
  • Shock resistant and leakproof
  • Smart choice for sampling and storing liquid, semi-liquid, and solid media
  • Sterile and food contact compliant
Often bought with…
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)100
  • MaterialHigh Density Polyethyene (HDPE)
  • Cap MaterialHDPE
  • Cap / Stopper Size37 mm
  • Cap ColorRed
  • Bottle ColorNatural
  • Special FeaturesRound
  • DescriptionRound Bottle, 100 mL, HDPE, 37 mm red screw cap, sterile; 145/cs
Key Features
  • Wide mouth with red screw cap
  • Shock resistant and leakproof
  • Smart choice for sampling and storing liquid, semi-liquid, and solid media
  • Sterile and food contact compliant
More About this Item
Full range of lightweight and leakproof HDPE storage bottles with an easy-access wide mouth. These bottles are easy to fill and empty out its contents. Bottles are fully assembled with a red screw cap for a leakproof seal. Conveniently sterilized to avoid any cross-contamination before final use.