Corning CoolRack® M30-PF Cooling Block, 30 x 500 µL Conical-Bottom Micrcentrifuge Tubes; Gray
Representative image only.
Corning Mfr # 432046 – Item # WW-04392-46
Cool, freeze, heat, or thaw samples uniformly
  • Place blocks directly into a temperature source from -196°C to over +100°C!
  • Obtain temperature uniformity of ±0.1°C across tubes and plates
  • Quickly reach bath temps—thermoconductive metal construction
  • 1 year warranty
PRODUCT OPTIONS
₹96,600.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Tubes Held30
  • ColorGray
  • Capacity30 x 500 µL conical-bottom tubes
  • DescriptionCooling Block, 30 x 500 µL Conical-Bottom Micrcentrifuge Tubes; Gray
  • Warranty1 year
1 Year Warranty
Key Features
  • Place blocks directly into a temperature source from -196°C to over +100°C!
  • Obtain temperature uniformity of ±0.1°C across tubes and plates
  • Quickly reach bath temps—thermoconductive metal construction
  • 1 year warranty
More About this Item
These aluminum blocks provide better temperature uniformity than placing samples directly into a bath. Aluminum blocks may be autoclaved, high-heat sterilized, or chemically treated to decrease cross-contamination concerns. Safe for use in ice, dry ice, alcohol, liquid nitrogen, water, oil, and other temperature regulation baths. Use in traditional ice buckets and pans, or use with a Corning CoolBox™ for an ice-free option.