Corning 430320 150 mL tube top vacuum filters, 0.22 µm, 12/cs
Representative image only.
Corning Mfr # 430320 – Item # WW-01385-62
Minimizes unnecessary transfers by filtering directly into centrifuge tube
  • Each polypropylene centrifuge tube is supplied with an individually wrapped cap for storage
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialCellulose Acetate
  • Pore Size (µm)0.22
  • Housing MaterialPolypropylene (PP)
  • Capacity (mL)150
  • Description150 mL tube top vacuum filters, 0.22 µm, 12/cs
Nonpyrogenic
Key Features
  • Each polypropylene centrifuge tube is supplied with an individually wrapped cap for storage
More About this Item
These filters feature 50 mm diameter Cellulose acetate (CA) membrane. Filters are individually packaged, sterile, certified nonpyrogenic. Each case includes two centrifuge tube stands.