Corning 96-well Microplate Lid, Low Evaporation Lid with Corner Notch, Sterile, Polystyrene; 50/Cs
Representative image only.
Corning Mfr # 3931 – Item # WW-01730-02
Designed to reduce contamination
  • Constructed of rigid polystyrene
  • Sterilized by gamma radiation
  • Packaged 25 lids per pack and 50 lids per case
PRODUCT OPTIONS
Call For Price
Often Bought With…
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells96
  • SterileYes
  • Plate Type96-Well Microplate Lid
  • Plate ColorClear
  • MaterialPolystyrene
  • Height (in)11/32
  • Width (in)3 23/64
  • Length (in)5 1/50
  • Packaging25 lids per pack and 50 lids per case
  • Description96-well Microplate Lid, Low Evaporation Lid with Corner Notch, Sterile, Polystyrene; 50/Cs
Key Features
  • Constructed of rigid polystyrene
  • Sterilized by gamma radiation
  • Packaged 25 lids per pack and 50 lids per case
More About this Item
The clear polystyrene (PS) lids are well suited for the Corning ® 96-well plates. Lids feature a corner-notch design with condensation rings to reduce contamination.