Corning 3789 Plate,round,white,100/cs, (White)
Representative image only.
Corning Mfr # 3789A – Item # WW-01728-60
  • Tested on a lot-to-lot basis for opacity and low background fluorescence/luminescence—assures minimal well-to-well crosstalk
  • Choose Black Polystyrene plates for minimizing background fluorescence and backlight scatter.
  • Well volume is 360 µL.
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells96
  • Well Volume (µL)360
  • Well ShapeRound
  • Surface TreatmentUntreated
  • Plate ColorWhite
  • DescriptionPlate,round,white,100/cs, (White)
Key Features
  • Tested on a lot-to-lot basis for opacity and low background fluorescence/luminescence—assures minimal well-to-well crosstalk
  • Choose Black Polystyrene plates for minimizing background fluorescence and backlight scatter.
  • Well volume is 360 µL.