Costar Cell Culture Multiwell Plate with Lid, Sterile, Ultra-Low Attachment Treated, 6-Well, 9.5 cm2; 24/Cs

Representative image only.
Costar Mfr # 3471 – Item # WW-01835-24
Unique hydrogel surface inhibits cell attachment
  • Reduce contamination with non-reversible lids with condensation ring
  • Individual alphanumeric codes for well identification
  • Sterilized by gamma radiation and nonpyrogenic
  • Individually wrapped to eliminate cross contamination

PRODUCT OPTIONS

Call For Price

SPECS & DESCRIPTION

Specifications & Description

  • Number of Wells6
  • Well Volume (mL)16.8
  • Well Bottom Diameter (mm)34.8
  • Well Top Diameter (mm)35.4
  • Well ShapeFlat
  • Culture Area (cm²)9.5
  • Surface TreatmentUltra-Low Attachment
  • Lid (Included)Yes
  • SterileYes
  • Plate ColorClear
  • MaterialPolystyrene
  • Height (in)29/32
  • Width (in)3 23/64
  • Length (in)5 1/32
  • Packaging1 Plate/Pack, 24 Plates/Case
  • DescriptionCell Culture Multiwell Plate with Lid, Sterile, Ultra-Low Attachment Treated, 6-Well, 9.5 cm2; 24/Cs

Key Features

  • Reduce contamination with non-reversible lids with condensation ring
  • Individual alphanumeric codes for well identification
  • Sterilized by gamma radiation and nonpyrogenic
  • Individually wrapped to eliminate cross contamination

More About this Item

These multiwell tissue culture plates have a unique covalently bonded hydrogel surface that is hydrophilc and neutrally charged. It minimizes cell attachment, protein absorption and enzyme activation. The surface is non-cytotoxic, biologically inert, and nondegradable. Use these products to maintain cells in a suspended, unattached state, prevent stem cells from attachment-mediated differentiation, prevent anchorage-dependent cells from dividing, and reduce binding of attachment and serum proteins to the substrate.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat