Corning Transwell® Permeable Supports, Sterile, PET Membrane, 0.4 µm, 12-Cluster Plate; 48/Cs

Representative image only.
Corning Mfr # 3460 – Item # WW-19480-15
Essential tool for the study of both anchorage-dependent and independent cell lines
  • Provide an environment for cells that closely resembles their in vivo state
  • Treated for optimal cell attachment
  • Ideal for a variety of cell culture applications—chemotaxis, drug transport, tissue remodeling, and invasion studies
  • Supports come sterile and preloaded in cluster plates
  • Sterilized by gamma radiation
  • Packaged 12 inserts in a 12 well plate, 4 plates per case, for a total of 48 inserts per case

PRODUCT OPTIONS

Call For Price

SPECS & DESCRIPTION

Specifications & Description

  • Cluster Plate Size Number12
  • Well ShapeFlat
  • Culture Area (cm²)1.12
  • Surface TreatmentTC-Treated
  • Lid (Included)Yes
  • SterileYes
  • Membrane MaterialPolyethylene Terephthalate (PET)
  • Membrane Diameter (mm)12
  • Height (mm)22.63
  • Length (mm)127.76
  • Width (mm)85.47
  • ColorClear
  • Pore Size (µm)0.4
  • DescriptionPermeable Supports, Sterile, PET Membrane, 0.4 µm, 12-Cluster Plate; 48/Cs

Key Features

  • Provide an environment for cells that closely resembles their in vivo state
  • Treated for optimal cell attachment
  • Ideal for a variety of cell culture applications—chemotaxis, drug transport, tissue remodeling, and invasion studies
  • Supports come sterile and preloaded in cluster plates
  • Sterilized by gamma radiation
  • Packaged 12 inserts in a 12 well plate, 4 plates per case, for a total of 48 inserts per case

More About this Item

These permeable supports, also known as cell culture inserts, are designed to satisfy your diverse cell culture requirements. A suspended design prevents media wicking between the insert and the plate well. It also allows for co-culturing without damaging cells attached to the plate's growth surface. Choose either polycarbonate (PC) or Polyethylene Terephthalate (PET) membrane material. Polycarbonate membranes are translucent while PET membranes are clear, allowing for excellent visibility under phase contrast microscopy.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat