Corning 3022P-25 Graduated Cylinder 25 mL PP, 2/pk
Representative image only.
Corning Mfr # 3022P-25 – Item # WW-06137-03
  • Meet ISO Standard 6706
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Capacity (mL)25
  • Graduation Interval (mL)0.5
  • DescriptionGraduated Cylinder 25 mL PP, 2/pk
Key Features
  • Meet ISO Standard 6706
More About this Item
Manufactured of translucent polpypropylene, these sturdy cylinders feature raised graduations and a wide hexagonal base. Calibrated "to contain" at 20°C. Autoclavable; exposure to temperatures above 140°F (60°C) may affect accuracy.