Corning 2159-24 Solvent Recovery Head, 1/Pk
Representative image only.
Corning Mfr # 2159-24 – Item # WW-34511-00
Use with Kuderna-Danish concentrators
  • Autoclavable
  • 24/40 Standard Taper Inner Joints
SPECS & DESCRIPTION
Specifications & Description
  • Accessory TypeSolvent Recovery Head
  • DescriptionSolvent Recovery Head, 1/Pk
Key Features
  • Autoclavable
  • 24/40 Standard Taper Inner Joints
More About this Item
Solvent recovery head is used for the condensation and collection of solvents. Use with Kuderna-Danish concentrator 34530-01 or -03. Length: 150 mm.