Corning 2158-1L Concentrator with Springs, 1000 Ml
Representative image only.
Corning Mfr # 2158-1L – Item # WW-34530-02
Efficiently boil away pestistides for accurate testing
  • Held together using glass hooks and two 1 1/4 inch stainless steel springs
SPECS & DESCRIPTION
Specifications & Description
  • Accessory TypeConcentrator
  • Range1000 mL
  • DescriptionConcentrator with Springs, 1000 Ml
Key Features
  • Held together using glass hooks and two 1 1/4 inch stainless steel springs
More About this Item
Kuderna-Danish Concentrators boil away samples to allow testing for pesticides or other pollutants prior to instrumental analysis. Include three-ball Snyder column with venting dimples and graduated collection tube. Available with springs mounted on collection tube.