CoorsTek 60371 99.5% Alumina Pestle, 100 mm Length; 1/Pk
Representative image only.
CoorsTek Mfr # 60371 – Item # WW-17823-28
SPECS & DESCRIPTION
Specifications & Description
  • Pestles Length (mm)100
  • Description99.5% Alumina Pestle, 100 mm Length; 1/Pk
More About this Item
Ideal for precise elemental analysis where minimal sample contamination is required. Pestles are glazed all over except for grinding surfaces. Mortar and pestle are sold as separate pieces. Autoclavable.