CoorsTek 60359 99.5% Alumina Pestle, 57 mm Length; 1/Pk
Representative image only.
CoorsTek Mfr # 60359 – Item # WW-17823-02
SPECS & DESCRIPTION
Specifications & Description
  • Pestles Length (mm)57
  • Description99.5% Alumina Pestle, 57 mm Length; 1/Pk
More About this Item
Ideal for precise elemental analysis where minimal sample contamination is required. Pestles are glazed all over except for grinding surfaces. Mortar and pestle are sold as separate pieces. Autoclavable.