This item is now discontinued. We recommend the following as suggested replacements: WW-17829-21

CoorsTek 66166 Porcelain Unglazed Rose Crucible, 75 mL; 6/Pk

Representative image only.
CoorsTek Mfr # 66166 – Item # WW-17809-00

SPECS & DESCRIPTION

Specifications & Description

  • MaterialPorcelain
  • Capacity (mL)75
  • Top Outside Diameter (mm)50
  • AutoclavableYes
  • DescriptionPorcelain Unglazed Rose Crucible, 75 mL; 6/Pk

More About this Item

Plain crucible without glaze. Ideal for general drying applications. Autoclavable.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat