Cooper-Atkins 35340 Aquatuff Datalogging Thermocouple Meter; Its, Probe
Representative image only.
Cooper-Atkins Mfr # 35340 – Item # WW-90025-18
Intelligent Temperature Stabilization (ITS) mode displays only the final stabilized temperature, eliminating any guesswork
  • Store up to 250 readings in memory
  • Fast-response probe with heavy-duty cable that is reinforced with aramid fiber
  • 5 year warranty
₹92,100.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Length (in)7.125
  • Height (in)1.2968
  • Width (in)2.75
  • DescriptionAquatuff Datalogging Thermocouple Meter; Its, Probe
  • Warranty5 years
5 Year WarrantyConformite Europeenne
Key Features
  • Store up to 250 readings in memory
  • Fast-response probe with heavy-duty cable that is reinforced with aramid fiber
  • 5 year warranty
More About this Item
This AquaTuff thermometer is waterproof and has Intelligent Temperature Stabilization (ITS), preventing the temperature from being displayed until stabilized. While in the ITS mode, you have the option of recording the stabilized temperature into memory. The memory can store up to 250 readings, which can then be reviewed by scrolling. When ITS mode is not selected, the instrument functions as a standard thermocouple thermometer.
The ABS housing is ergonomically designed with a large easy-to-read display and a curved back that fits nicely in your hand. Powerful microprocessor delivers speed and reliability with a unique memory that stores the calibration settings and reduces the need for frequent recalibration.
Other features include selectable °F/°C units, user selectable resolutions, Hold function, auto-off, and watertight battery hatch.