Contec EasySat Mop Head for Bucketless Floor Mop
Representative image only.
Contec Mfr # BLVKMOP – Item # WW-09411-31
₹70,800.00INR / Pkg of 32
SPECS & DESCRIPTION
Specifications & Description
  • MaterialUltraclean polyester/ester foam blend
  • DescriptionMop Head for Bucketless Floor Mop
More About this Item
EasySat™ Mop heads are made from ultra-clean polyester/ester foam VertiKlean material. Available with flat or textured surface and standard or gamma irradiated to 10-6 Sterility Assurance Level per AAMI/ISO 11137:2006 guidelines.