Contec EasyReach Cleaning Tool, 17" length
Representative image only.
Contec Mfr # 2665SF – Item # WW-09411-15
Long handle reduces the risk of operators leaning into areas and possibly introducing contaminates
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless steel
  • Handle Length (in)17
  • Width (in)7 1/2
  • Length (in)17
  • DescriptionCleaning Tool, 17\" length
ISO9001 Certified Supplier
More About this Item
Prevent the risk of operators introducing contaminates into a system by keeping the operator out! Ideal for use when cleaning laminar airflow hoods, restricted access barrier systems, isolators, biosafety cabinets, glove boxes, and more! The 17" (43 cm) handle extends your reach while the pivoting head making cleaning those hard-to-reach areas easier. Stainless steel tool is autoclavable.
Quilted polyester cleaning pads (sold separately) feature a tab/grip that easily fits onto the tool even while you''re wearing gloves. Choose from dry or wet and standard or sterile pads.