Conductivity Probe, Glass Body, K=1.0
Representative image only.
Item # WW-19601-26
Optimize performance of your conductivity and pH/ORP meters
  • Get accurate, reliable measurements with these replacement probes and adapters
  • Features a conductivity range of 0.05 to 200,000 microsiemens
  • Suitable for use with 19601-06 meter
  • 1 year warranty
PRODUCT OPTIONS
₹28,100.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • Compatible WithTraceable 19601-06 conductivity meter
  • DescriptionConductivity Probe, Glass Body, K=1.0
  • Warranty1 Year
1 Year WarrantyMade in the U.S.A.
Key Features
  • Get accurate, reliable measurements with these replacement probes and adapters
  • Features a conductivity range of 0.05 to 200,000 microsiemens
  • Suitable for use with 19601-06 meter
  • 1 year warranty
More About this Item
Replace or upgrade electrodes, probes, cells, and parts for Traceable meters.