Cole-Parmer Waste System, UN/DOT, 70 mm Cap, 6 x 1/8" OD Tubing; 60 L
Representative image only.
Cole-Parmer Item # WW-06062-64
Provide safe HPLC waste disposal in your lab
  • Certified for UN/DOT Packaging Group II for hazardous chemicals
  • Carbon filter captures potentially harmful vapors
  • Multiple ports adapt cap to any HPLC system
  • Filter adapter elbow provides storage flexibility, especially for tight lab spaces
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (L)60
  • Capacity (Gal)15.85
  • MaterialHigh Density Polyethyene (HDPE)
  • Cap MaterialPolypropylene (PP)
  • Cap / Stopper Size70 mm
  • Cap ColorWhite
  • Bottle ColorWhite
  • Special Features6 x 1/8\" OD Tubing
  • AutoclavableNo
  • DescriptionWaste System, UN/DOT, 70 mm Cap, 6 x 1/8\" OD Tubing; 60 L
Key Features
  • Certified for UN/DOT Packaging Group II for hazardous chemicals
  • Carbon filter captures potentially harmful vapors
  • Multiple ports adapt cap to any HPLC system
  • Filter adapter elbow provides storage flexibility, especially for tight lab spaces
More About this Item
This solvent waste system allows easy and safe disposal of HPLC waste in the lab. The system includes a UN/DOT container with two caps; one with ports for waste collection and one to seal for transportation. Caps come with a variety of port sizes to be used with complex HPLC systems. When not in use, the ports can be plugged.