Foxx Life Sciences Waste System Cap w/ Exhaust Filter, 4 x 1/8" and 3 x 1/4" OD tubing
Representative image only.
Mfr # 330-0413-OEM – Item # WW-06063-21
Protect your lab from harmful solvent vapors
  • Carbon exhaust filter traps fumes
  • Tubing attaches securely to prevent escaping vapors and spills
  • Elbow adapter included to save on vertical space
₹52,700.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Cap MaterialPolypropylene
  • Cap / Stopper Size83 mm
  • Cap ColorWhite
  • Special Features4 x 1/8\" & 3 x 1/4\" OD tubing
  • DescriptionWaste System Cap w/ Exhaust Filter, 4 x 1/8\" and 3 x 1/4\" OD tubing
Key Features
  • Carbon exhaust filter traps fumes
  • Tubing attaches securely to prevent escaping vapors and spills
  • Elbow adapter included to save on vertical space
More About this Item
These carboy cap assemblies feature tubing ports with fittings to ensure a leak-tight connection and an activated carbon exhaust filter to remove solvent fumes from escaping air. Ideally suited for HPLC waste collection, the cap is constructed of polypropylene and includes a replaceable sulfur-cured EPDM gasket. Tubing fittings are made of acetal and ETFE for compatibility with HPLC-grade solvents. Caps fit standard carboys and jerricans with 83-mm diameter neck.