Cole-Parmer Vortex Mixer Rack, 9 x 50 mL Tubes (horizontal; QuEChERS); 1/Ea
Representative image only.
Cole-Parmer Item # WW-04725-68
Pre-drilled foam racks to meet your mixing needs
  • Use with Cole-Parmer High-Capacity Vortex Mixer/Shaker—standard or QuEChERS versions.
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Compatible With9 x 50 mL tubes (horizontal; QuEChERS)
  • DescriptionVortex Mixer Rack, 9 x 50 mL Tubes (horizontal; QuEChERS); 1/Ea
  • Warranty2 Years
Two year warranty
Key Features
  • Use with Cole-Parmer High-Capacity Vortex Mixer/Shaker—standard or QuEChERS versions.
  • 2 year warranty
More About this Item
Lightweight foam racks can be used to hold samples vertically or horizontally.