Cole-Parmer Essentials Versatile Carboy Cap, 83B, 2 x 1/2" ID Ports; 1 Ea
Representative image only.
Cole-Parmer Essentials Item # WW-06059-27
Leakproof caps with the flexibility of interchangeable ports
  • Finger grips allow easy tightening and opening
  • Choose from a variety of sizes and configurations*
  • Change the top connections of each carboy to match your application needs
  • Autoclavable
SPECS & DESCRIPTION
Specifications & Description
  • Cap MaterialPP
  • Cap / Stopper Size83B
  • Cap ColorWhite
  • AutoclavableYes
  • Tubing ID2 x 1/2\" ID Hosebarb Ports
  • DescriptionVersatile Carboy Cap, 83B, 2 x 1/2\" ID Ports; 1 Ea
Key Features
  • Finger grips allow easy tightening and opening
  • Choose from a variety of sizes and configurations*
  • Change the top connections of each carboy to match your application needs
  • Autoclavable
More About this Item
Versatile carboy caps are made of autoclavable polypropylene and come in industry-standard sizes.
Solid caps do not accept insert adapters.