Cole-Parmer VapLock™ Fitting, Brown PEEK™ w/ White PCTFE, Straight, Compression to Threaded Adapter, 4.0 mm OD x 5/16-24 UNF(M); 1/EA
Representative image only.
Cole-Parmer Item # WW-12020-01
Designed for secure liquid-tight connections
  • Two piece nut and ferrule system
  • Adapter for semi-rigid tubing
  • Color coded for your convenience
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPEEK / PCTFE
  • Connection TypeCompression x UNF(M)
  • Thread size UNF(M) (port 2) (in)5/16-24
  • Compression Size / Tube Od (Port 1) (in)5/32
  • Compression Size / Tube Od (Port 1) (mm)4
  • ColorBrown / White
  • DescriptionTubing Adapter, 4.0 mm OD x 5/16-24 UNF(M), brown PEEK™ w/ white PCTFE; 1/EA
Key Features
  • Two piece nut and ferrule system
  • Adapter for semi-rigid tubing
  • Color coded for your convenience
More About this Item
Adapters for semi-rigid tubing are color coded by tubing size for easy identification. A two-piece nut and ferrule system provides a liquid-tight connection and withstands pressures up to 500 psi.