Cole-Parmer VapLock™ Fitting, Blue PP Nut, Yellow ETFE Ferrule, Straight, Compression to Threaded Adapter, 1/8" OD x 1/4" NPT(M); 1/EA
Representative image only.
Cole-Parmer Item # WW-12020-16
Designed for secure leak-resistant connections
  • Easily identify from a distance with color coded adapters
  • 1/8" OD x 1/4" NPT(M)
  • Blue PP nut and yellow ETFE ferrule
PRODUCT OPTIONS
₹2,400.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPP / ETFE
  • Connection TypeCompression x NPT(M)
  • Thread size NPT(M) (port 2) (in)1/4
  • Compression Size / Tube Od (Port 1) (in)1/8
  • Compression Size / Tube Od (Port 1) (mm)3.175
  • ColorBlue nut/ yellow ferrule/white adapter
  • Max Operating Pressure (PSI)500
  • DescriptionTubing Adapter, 1/8\" OD x 1/4\" NPT(M), Blue PP Nut, Yellow ETFE Ferrule; 1/EA
Key Features
  • Easily identify from a distance with color coded adapters
  • 1/8" OD x 1/4" NPT(M)
  • Blue PP nut and yellow ETFE ferrule
More About this Item
Adapters for semi-rigid tubing are color coded by tubing size for easy identification. Each connection is comprised of a two-piece nut and ferrule system with polypropylene NPT(M) thread adapter. Wrap NPT thread 3 to 4 times with PTFE tape and wrench tighten for a leak-resistant connection. Adapters withstand pressures up to 500 psi.