Cole-Parmer VapLock™ Solvent Delivery Cap, two 1/4"-28 ports, 38 mm Wako; 1/ea

Representative image only.
Cole-Parmer Item # WW-12018-39
Reduce chemical evaporation and exposure to hazardous vapors
  • Use with semi-rigid tubing equal to or less than 1/8" (3.2 mm) OD

PRODUCT OPTIONS

₹5,000.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Cap MaterialPTFE/PP
  • Cap / Stopper SizeWako 38 mm
  • Cap ColorWhite
  • Special Features2 x 1/4-28 UNF(F)
  • AutoclavableYes
  • DescriptionSolvent Delivery Cap, two 1/4-28 ports, 38 mm Wako

Key Features

  • Use with semi-rigid tubing equal to or less than 1/8" (3.2 mm) OD

More About this Item

Create a closed system using solvent reservoir caps to prevent hazardous vapors from escaping into the lab. Caps accept standard 1/4"-28 UNF(M) fittings to easily connect semi-rigid tubing. Anti-twist design allows the inner body of the cap to rotate independently preventing tubes from tangling when caps are removed or attached. Caps are constructed of PTFE body and PP collar* and are autoclavable.

Caps with threaded port have ports angled at 10° tilt for more accessible tubing connections. Select caps with stainless steel (SS) thread insert to minimize cross-threading and damage to soft PTFE ports.

*Does not come in contact with liquid under normal circumstances.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat