Cole-Parmer VapLock™ Luer Fitting, PVDF, Plug, Male Luer Lock, Closed at Grip; 1/EA
Representative image only.
Cole-Parmer Mfr # MTLLP-J1A – Item # WW-12028-60
Quick connection and release make these luer fittings excellent for many applications
  • Great for connecting to syringes and small tubing
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyvinylidene fluoride (PVDF)
  • Number of Ports2
  • Connection Type-Port 1Male luer lock
  • Connection Type-Port 2Plug, closed at grip
  • DescriptionLuer Fitting, PVDF, Plug, Male Luer Lock, Closed at Grip; 1/EA
Key Features
  • Great for connecting to syringes and small tubing
More About this Item
Choose from fittings constructed from durable polypropylene (PP) or chemically inert PVDF.