Cole-Parmer VapLock™ Breakthrough Indicator for Exhaust Filters; 5/pk
Representative image only.
Cole-Parmer Item # WW-12019-83
Use with exhaust filters to detect end-of-service life
  • Compatible with Cole-Parmer® VapLock™ activated carbon exhaust filters
PRODUCT OPTIONS
₹29,500.00INR / Pkg of 5
SPECS & DESCRIPTION
Specifications & Description
  • MaterialAcrylic
  • Compatible WithVapLock™ activated carbon exhaust filters
  • DescriptionBreakthrough Indicator for Exhaust Filters; 5/pk
Key Features
  • Compatible with Cole-Parmer® VapLock™ activated carbon exhaust filters
More About this Item
Color change occurs when the indicator is exposed to organic solvent vapors. Indicator housing is made of acrylic.