Spex VapLock™ Fitting, Polypropylene, Straight, Hose Barb Union, 3/8" ID; 10/PK
Representative image only.
Spex Item # WW-12020-89
Connect soft tubing using barbed fittings
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Straight fitting for 3/8" ID tubing
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolypropylene
  • Number Of Ports2
  • Barb Size / Tubing Id (Port 1) (in)3/8
  • Barb Size / Tubing Id (Port 2) (in)3/8
  • ColorTranslucent
  • DescriptionFitting, Polypropylene, Straight, Hose Barb Union, 3/8\" ID; 10/PK
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Straight fitting for 3/8" ID tubing
More About this Item
These straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.